Learn More
Abcam Ca2+/Calmodulin Solution, 10x Stock, 500 μL
Description
- 10x stock solution
- Used for assaying protein kinase activity of CAMK family members
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Blue Ice |
| Target Molecule | Ca2+, Calmodulin |
| Form | Solution |
| Content And Storage | Store at -20°C. |
| Format | Vial |
| Quantity | 500 μL |
| For Use With (Application) | Protein kinase activity assay |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.