Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAB13 Rabbit anti-Plant, Polyclonal, FabGennix

Catalog No. 502206315
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6315 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6315 Supplier Fabgennix Inc Supplier No. CAB13101AP
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CAB13 Polyclonal antibody specifically detects CAB13 in Plant samples. It is validated for Dot Blot, ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen CAB13
Applications Dot Blot, ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene Alias CAB13; CAB13 LHBC1; Chlorophyll a-b binding protein 13,chloroplastic
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 100-200 on Solanum lycopersicum Chlorophyll a-b binding protein 13.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Plant
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.