Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Cabergoline, D2-Like Receptor Agonist, >99% Purity, Solid, 10 mg

Catalog No. 502707481
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7481 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7481 Supplier Abcam Supplier No. AB12056410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent D2-like receptor agonist with high affinity for selected 5-HT receptors.

  • Molecular weight: 451.6 g/mol
  • Purity: >99%
  • Form: Solid
  • Activity: D2-like receptor agonist (Ki values are 0.7 (D2), 1.5 (D3), 9.0 (D4) and 165 nM (D5)); potent at some 5-HT receptors (Ki values are 20 (5-HT1A), 8.7 (5-HT1D), 6.2 (5-HT2A) and 1.2 nM (5-HT2B))
  • Additional properties: Shows antitumour effects, antidepressant and anxiolytic properties
  • Storage: Store at +4°C
  • Shipping: Ambient - Can Ship with Ice

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Buffer 100% Cabergoline
Content And Storage Store at 4°C.
Form Solid
Shipping Condition Ambient
Target Molecule D2-like Receptors, 5-HT Receptors
Quantity 10 mg
For Use With (Application) D2-like receptor agonism, 5-HT receptor research, antitumour research, antidepressant research, anxiolytic research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.