Learn More
Abcam Cabergoline, D2-Like Receptor Agonist, Solid, >99% Purity, 50 mg
Description
- Purity: >99%
- Molecular Weight: 451.6 g/mol
- Form: Solid
- Activity: D2-like receptor agonist (Ki values: 0.7 (D2), 1.5 (D3), 9.0 (D4), 165 nM (D5))
- Additional Pharmacology: Highly potent at select 5-HT receptors (Ki values: 20 (5-HT1A), 8.7 (5-HT1D), 6.2 (5-HT2A), 1.2 nM (5-HT2B))
- Storage: Store at +4°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Content And Storage | Store at 4°C. |
| Form | Solid |
| Shipping Condition | Ambient |
| Target Molecule | D2-like receptor, 5-HT receptor |
| Formulation | 100% Cabergoline |
| Quantity | 50 mg |
| For Use With (Application) | D2-like receptor agonist, 5-HT receptor modulation |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.