Description
- Slotted tip
- Fluted grip
- Beryllium copper material
- Lifetime warranty
- Made in the USA
Specifications
Specifications
| Product Type | Cabinet-Tip Screwdriver |
| Length (English) | 6 -5/8 in. |
| Tip Style | 3/16 in. Slotted |