Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CABLES2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599599
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599599 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599599 Supplier Invitrogen™ Supplier No. PA599599
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Cables2.

Unknown. Probably involved in G1-S cell cycle transition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CABLES2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CABLES2
Gene Accession No. Q8K3M5, Q9BTV7
Gene Alias C20orf150; Cables2; Cdk5 and Abl enzyme substrate 2; CDK5 and ABL1 enzyme substrate 2; dJ908M14.2; ik3-2; Interactor with CDK3 2
Gene Symbols CABLES2
Host Species Rabbit
Immunogen A synthesized peptide derived from human CABLES2(Accession Q9BTV7), corresponding to amino acid residues P94-Q144.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 252966, 311703, 81928
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.