Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CACNA2D1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01674821
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-821 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-821 Supplier Invitrogen™ Supplier No. PA112667
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control for this product is rat brain lysate.

This gene encodes a member of the alpha-2/delta subunit family, a protein in the voltage-dependent calcium channel complex. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization and consist of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. Research on a highly similar protein in rabbit suggests the protein described in this record is cleaved into alpha-2 and delta subunits. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CACNA2D1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene Cacna2d1
Gene Accession No. P54289, P54290
Gene Alias Ca(v)alpha2delta1; Cacna2; Cacna2d1; CACNL2A; Calcium channel subunit alpha 2 delta (dihydropyridine - sensitive L-type); calcium channel, L type, alpha 2 polypeptide; calcium channel, voltage-dependent, alpha 2/delta subunit 1; calcium channel, voltage-dependent, alpha2/delta subunit 1; calcium voltage-gated channel auxiliary subunit alpha2delta 1; CCHL2A; CCHLA2; DHP; DHSCCA; dihydropryridine-sensitive calcium channel alpha-2 subunit; dihydropyridine-sensitive calcium channel alpha 2; dihydropyridine-sensitive L-type, calcium channel alpha-2/delta subunit; H_DJ0560O14.1; LINC01112; lncRNA-N3; MHS3; unknown protein; voltage-dependent calcium channel alpha-2 delta subunit precursor; Voltage-dependent calcium channel subunit alpha-2/delta-1; Voltage-dependent calcium channel subunit alpha-2-1; Voltage-dependent calcium channel subunit delta-1; Voltage-gated calcium channel subunit alpha-2/delta-1; WUGSC:H_DJ0560O14.1
Gene Symbols Cacna2d1
Host Species Rabbit
Immunogen Synthetic peptide, conjugated to KLH, corresponding to the rat alpha2/delta calcium channel.
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25399, 781
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.