Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CACNB1 Monoclonal Antibody (OTI3H6), TrueMAB™, OriGene
SDP

Catalog No. 501682749
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-2749 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-2749 Supplier OriGene Supplier No. TA809369
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The expression of multiple classes of voltage dependent calcium channels (VDCC′s) allows neurons to tailor calcium signaling to functionally discrete cellular regions. Although N-Type VDCC′s exist before birth which is consistent with a role in migration, most N-Type VDCC′s subunit expression is postnatal and is important in the genesis of synaptic transmission in discrete hippocampal fields.

Specifications

Antigen CACNB1
Applications Western Blot
Classification Monoclonal
Clone OTI3H6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CACNB1
Gene Accession No. Q02641
Gene Alias CAB1, CACNLB1, CCHLB1
Gene Symbols CACNB1
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-269 of human CACNB1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 782
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.