Learn More
Invitrogen™ CACNB2 Polyclonal Antibody

Description
A suggested positive control for this product is rat brain lysate.
Specifications
Specifications
| Antigen | CACNB2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene | Cacnb2 |
| Gene Accession No. | Q08289 |
| Gene Alias | AW060387; brain calcium channel beta-subunit CaB2c (splice variant of the gene CaB2); CAB1; CAB2; Cacnb2; CACNLB1; Cacnlb2; Calcium channel voltage-dependent subunit beta 2; calcium channel, voltage-dependent, beta 2 subunit; calcium voltage-gated channel auxiliary subunit beta 2; cardiac calcium channel beta subunit; cardiac calcium channel beta-subunit CaB2a (splice variant of the gene CaB2; cardiac calcium channel beta-subunit CaB2b (splice variant of the gene CaB2); CAVB2; Cavbeta2; Cchb2; CCHLB1; lambert-Eaton myasthenic syndrome antigen B; L-type calcium channel beta subunit; L-type voltage-dependent calcium channel beta-2 subunit; MGC138502; MGC138504; MGC41896; myasthenic (Lambert-Eaton) syndrome antigen B; MYSB; voltage-dependent calcium channel beta2A subunit; voltage-dependent calcium channel beta2B subunit; voltage-dependent L-type calcium channel subunit beta-2; voltage-gated calcium channel beta2 subunit transcript variant X13 |
| Gene Symbols | Cacnb2 |
| Host Species | Rabbit |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.