Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CACNB3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589997
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589997 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589997 Supplier Invitrogen™ Supplier No. PA589997
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VYWRATHHPA PGPGLLGPPS AIPGLQNQQL LGERGEEHSP LERDSLMPSD EASESSRQAW TGSSQRSSRH LEEDYADAYQ DLYQPHRQHT SGLPSANGHD PQDRLLAQDS EHNHSDRNWQ RNRPWPKDSY; Positive Samples: Mouse brain, Rat brain.

CACNB3 encodes a regulatory beta subunit of the voltage-dependent calcium channel. Beta subunits are composed of five domains, which contribute to the regulation of surface expression and gating of calcium channels and may also play a role in the regulation of transcription factors and calcium transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CACNB3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.03 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Cacnb3
Gene Accession No. P54285, P54287
Gene Alias Beta3; Ca(v)beta3; CAB3; CACH3B; Cacnb3; Cacnlb3; Calcium channel subunit beta 3; calcium channel voltage-dependent subunit beta 3; calcium channel, voltage-dependent, beta 3 subunit; calcium voltage-gated channel auxiliary subunit beta 3; cardiac calcium channel beta-subunit CaB3; Cav; CCHB3; voltage-dependent calcium channel beta3A subunit; voltage-dependent L-type calcium channel subunit beta-3
Gene Symbols Cacnb3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 355-484 of human CACNB3 (NP_0007162).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12297, 25297
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.