Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CACNG3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109290
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109290 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109290 Supplier Invitrogen™ Supplier No. PA5109290
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: SH-SY-5Y, rat brain tissue.

Calcium Channel gamma-3 is a member of voltage dependent calcium channels (VDCCs) which allows neurons to tailor calcium signaling to functionally discrete cellular regions. Although N-Type VDCCs exist before birth which is consistent with a role in migration, most N-Type VDCCs subunit expression is postnatal, and important in the genesis of synaptic transmission in discrete hippocampal fields. Calcium Channel gamma-3 is a type I transmembrane AMPA receptor regulatory protein (TARP). TARPs regulate both trafficking and channel gating of the AMPA receptors. Further, Calcium Channel gamma-3 is part of a functionally diverse eight-member protein subfamily of the PMP-22/EMP/MP20 family. Calcium Channel gamma-3 gene dysfunction is a susceptibility locus for childhood absence epilepsy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CACNG3
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene Cacng3
Gene Accession No. O60359, Q8VHX0
Gene Alias Cacng3; CACNLB3; calcium channel, voltage-dependent, gamma subunit 3; calcium voltage-gated channel auxiliary subunit gamma 3; CaVgamma3; neuronal voltage-gated calcium channel gamma-3 subunit; TARP gamma-3; transmembrane AMPAR regulatory protein gamma-3; Voltage-dependent calcium channel gamma-3 subunit; voltage-gated calcium channel gamma subunit
Gene Symbols Cacng3
Host Species Rabbit
Immunogen Synthetic peptide within rat CACNG3 aa 192-241.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10368, 140724
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.