Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CACNG7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109476
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109476 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109476 Supplier Thermo Scientific Supplier No. PA5109476
Only null left
Add to Cart
Add to Cart

CACNG7 Polyclonal Antibody

CACNG7 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen CACNG7
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.92 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Cacng7
Gene Accession No. P62955, P62956, P62957
Gene Alias Cacng7; calcium channel, voltage-dependent, gamma subunit 7; calcium voltage-gated channel auxiliary subunit gamma 7; neuronal voltage-gated calcium channel gamma-7 subunit; TARP gamma-7; transmembrane AMPAR regulatory protein gamma-7; voltage dependent calcium channel gamma 7 subunit; voltage-dependent calcium channel gamma-7 subunit
Gene Symbols Cacng7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 156-275 of human CACNG7 (NP_114102.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140728, 59284, 81904
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.