Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadence Science™ Micro-Emulsifying Needles
SDP

Catalog No. 1482517H Shop All Cadence Science Inc. Products
Change view
Click to view available options
Needle Gauge:
16 G
18 G
20 G
Includes:
Reinforcing bar
Without reinforcing bar
Length (English) Needle:
1.87 in.
2.87 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Includes Length (English) Needle
14-825-17H 20 G Reinforcing bar 2.87 in.
14-825-17A 16 G Reinforcing bar 2.87 in.
14-825-17J 18 G Reinforcing bar 2.87 in.
14-825-17L 18 G Without reinforcing bar 1.87 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-825-17H Supplier Cadence Science™ Supplier No. 7976
Add to Cart
Add to Cart

Double-hub needles are designed for use in mixing two liquid media differing in viscosity—especially for adjuvants and antigens, oil-based liquids, allergens

  • Available with or without a reinforcing bar
  • Overall length: 2 7/8 in. (7.3cm), except for 14-825-17K and -17L needles which have a length of 1 7/8 in. (4.8cm)

Specifications

Material Glass
Needle Gauge 20 G
For Use With (Application) For allergens and the mixing of adjuvants and antigens, oil based liquids or any two liquid media differing in viscosity, with reinforcing bar
Includes Reinforcing bar
Length (English) Needle 2.87 in.
Product Type Micro Emulsifying Needle with Reinforcing Bar
Length (Metric) Needle 7.3 cm
Sterility Non-sterile
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.