Learn More
Cadence Science™ Non-Coring Septum Penetration Needles
SDP
Catalog No. 0129024 Shop All Cadence Science Inc. Products
Change view
Click to view available options
Needle Gauge:
18 G
20 G
Length (English) Needle:
6 in.
12 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Length (English) Needle
01-290-24 20 G 6 in.
01-290-22 18 G 12 in.
01-290-25 18 G 6 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-290-24 Supplier Cadence Science™ Supplier No. 9863
Add to Cart
Add to Cart
Needles allow for easier penetration and minimize coring and tracking
  • Micro-mate 316 stainless-steel hub
  • 316 stainless-steel tubing
Specifications
Material Stainless Steel (Hub)
Needle Gauge 20 G
For Use With (Application) For easier penetration and minimize coring and tracking
Length (English) Needle 6 in.
Product Type Penetration Needle
Quantity 12 Needles
Needle Point Style Deflected End
WARNING: Cancer - www.P65Warnings.ca.gov