Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadherin 13/CDH13/T-cadherin Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501569152
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-9152 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-9152 Supplier Bethyl Laboratories, Inc Supplier No. A304967AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

The cadherins are a family of Ca++-dependent adhesion molecules that function to mediate cell-cell binding critical to the maintece of tissue structure and morphogenesis. Cadherins each contain a large extracellular domain at the amino terminus, which is characterized by a series of five homologous repeats, the most distal of which is thought to be responsible for binding specificity. The relatively short carboxy terminal, intracellular domain interacts with a variety of cytoplasmic proteins, including beta-catenin, to regulate cadherin function. T-cadherin (for truncated-cadherin, also designated heartcadherin or cadherin-13) expression levels have been shown to be reduced in human breast cancers and carcinoma cells lines. Evidence suggests that.

Specifications

Antigen Cadherin 13/CDH13/T-cadherin
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene CDH13
Gene Accession No. P55290
Gene Alias Cadherin-13; Truncated cadherin; T-cadherin; T-cad; Heart cadherin; H-cadherin; P105; CDH13; CDHH
Gene Symbols CDH13
Host Species Rabbit
Immunogen Between 475 and 525
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1012
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.