Learn More
Description
Host Species: Mouse
Clone: 56C1
Isotype: IgG1
Species Reactivity: Human. Others not known.
Epitope: External domain
Immunogen: Recombinant protein corresponding to extracellular domain of human cadherin-P
Molecular Weight: 120kDa
Positive Control: Skin
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: IVD; Int'l: IVD
Specifications
Specifications
| Antigen | Cadherin-P/P-cadherin Ab-1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 56C1 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant protein corresponding to extracellular domain of human P-cadherin |
| Purification Method | Skin |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.