Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CADM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532687
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532687 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532687 Supplier Invitrogen™ Supplier No. PA532687
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Bovine, Dog, Bat, Elephant, Panda, Horse, Rabbit, Pig, Opossum (100%).

This gene encodes a member of the synaptic cell adhesion molecule 1 (SynCAM) family which belongs to the immunoglobulin (Ig) superfamily. The encoded protein has three Ig-like domains and a cytosolic protein 4.1 binding site near the C-terminus. Proteins belonging to the protein 4.1 family crosslink spectrin and interact with other cytoskeletal proteins. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CADM2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene CADM2
Gene Accession No. Q8N3J6
Gene Alias 2900078E11Rik; 9330131D06; A830029E02Rik; Cadm2; Cell adhesion molecule 2; Igdf4d; Igsf4d; immunoglobulin superfamily member 4D; immunoglobulin superfamily, member 4; immunoglobulin superfamily, member 4D; NECL3; Necl-3; nectin-like 3; nectin-like protein 3; Synaptic cell adhesion molecule 2; SynCAM 2; SynCAM2
Gene Symbols CADM2
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from extracellular domain of human CADM2.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100069765, 100340168, 100626294, 253559, 487680, 517759
Target Species Human, Canine, Rabbit, Monkey, Bovine, Pig, Horse, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.