Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium Acetate Dihydrate, purum p.a., ≥98.0% (KT), Solstice

Catalog No. 60045179 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-179 10 g Poly Bottle
60-045-180 500 g Poly Bottle
60-045-178 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-045-179 Supplier Solstice Supplier No. 2090110G

Purum p.a.,≥98.0%

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5743-04-4
Molecular Formula C4H10CdO6
Molecular Weight (g/mol) 266.53
MDL Number MFCD00150020
InChI Key AUIZLSZEDUYGDE-UHFFFAOYSA-L

Specifications

Melting Point 256°C
Physical Form Solid
Quantity 10 g
Linear Formula Cd(CH3COO)2 · 2H2O
UN Number UN3077
Beilstein 3730567
Formula Weight 266.53g/mol
Percent Purity ≥98.0% (KT)
Grade purum p.a.
Packaging Poly Bottle
Density 2.01g/cu.cm
Chemical Name or Material Cadmium Acetate Dihydrate
Show More Show Less
Hazard Statement P201-P261-P273-P280-P308 + P313-P501
Precautionary Statement H302 + H312 + H332-H350-H410
DOTInformation 9/PGIII
EINECSNumber 208-853-2
RTECSNumber AF7505000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.