Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium Chloride Hemi(Pentahydrate), 79.5 to 81.0%, ACS Reagent, Solstice

Catalog No. 60045483 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-483 100 g Poly Bottle
60-045-485 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-045-483 Supplier Solstice Supplier No. 239208100G

ACS Reagent, 79.5-81.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-78-5
MDL Number MFCD00799071

Specifications

Quantity 100 g
Linear Formula CdCl2·21/2H2O
UN Number UN2570
Formula Weight 228.36g/mol
Percent Purity 79.5 to 81.0%
Grade ACS Reagent
Packaging Poly Bottle
Vapor Pressure 10mmHg (656°C)
Density 3.327g/cm3; Vapor Density: 6.3 (vs air)
Chemical Name or Material Cadmium Chloride Hemi(Pentahydrate)
Hazard Statement P201-P260-P273-P284-P301 + P310 + P330-P304 + P340 + P310
Precautionary Statement H301-H330-H340-H350-H360FD-H372-H410
DOTInformation Class 6.1 - PG 3 - EHS - Cadmium compounds, HI: all (not BR)
EINECSNumber 233-296-7
RTECSNumber EV0178000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.