Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium iodide, 99%

Catalog No. AC211460010
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC211460010 1 kg Glass bottle
AC211462500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC211460010 Supplier Thermo Scientific Chemicals Supplier No. 211460010
Add to Cart

CAS: 7790-80-9 | CdI2 | 366.223 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-80-9
MDL Number MFCD00010920
InChI Key OKIIEJOIXGHUKX-UHFFFAOYSA-L
Synonym cadmium iodide, cadmium diiodide, unii-2f2upu4kcw, cadmium 2+ diiodide, 2f2upu4kcw, acmc-20aljc, cadmium 2+ ion diiodide, cadmium iodide trace metals basis 25g
PubChem CID 24635
IUPAC Name cadmium(2+);diiodide
SMILES [Cd+2].[I-].[I-]

Specifications

Boiling Point 787°C
Melting Point 388°C
Color White to Yellow
Physical Form Shiny Crystalline Flakes or Powder
Quantity 1 kg
Assay Percent Range 98.5% min. (Complexometry)
Molecular Formula CdI2
Linear Formula CdI2
Merck Index 15, 1626
Solubility Information Solubility in water: 798g/L (20°C)
Molecular Weight (g/mol) 366.2
Formula Weight 366.2
Percent Purity 99%
Packaging Glass bottle
Chemical Name or Material Cadmium iodide, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 232-223-6
RTECSNumber EV1290000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.