Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium Sulfate, 8/3-Hydrate, Crystal, Reagent, ACS, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18601180 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-180 25 g Amber Glass Bottle
18-601-181 125 g Poly Bottle
18-601-182 500 g Poly Bottle
18-601-183 12 kg Poly Pail
18-601-184 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-601-180 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C105025GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

C1050, 7790-84-3, CdSO4·8/3H2O

Spectrum™ Chemical Cadmium Sulfate, 8/3-Hydrate, Crystal, Reagent, ACS , is used widely for the electroplating of cadmium in electronic circuits. It is also a precursor to cadmium-based pigment such as cadmium sulfide. It is also used for electrolyte in a Weston standard cell as well as a pigment in fluorescent screens. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 7790-84-3
Molecular Formula CdH2O5S
Molecular Weight (g/mol) 226.49
InChI Key VRFRVKRTIMIRNG-UHFFFAOYSA-L
IUPAC Name cadmium(2+) hydrate sulfate
SMILES O.[Cd++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 100%
Quantity 25 g
UN Number UN2570
Percent Purity 98 to 102%
Grade ACS
Packaging Amber Glass Bottle
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.