Learn More
CPC Scientific Glp-Gln-Asp-Tyr(SO3H)-Thr-Gly-Trp-Met-Asp-Phe-NH2 1MG
SDP
Supplier:  CPC Scientific CCKS001B
Encompass
SEQUENCE: Glp-Gln-Asp-Tyr(SO3H)-Thr-Gly-Trp-Met-Asp-Phe-NH2ONE-LETTER SEQUENCE: Glp-QD-Tyr(SO3H)-TGWMDF-NH2MOLECULAR FORMULA: C58H73N13O21S2MOLECULAR WEIGHT: 1352.4STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [17650-98-5]SYNONYMS: Ceruletide, CeruleinRESEARCH AREA: GastrointestinalREFERENCES:A. Anastasi et al., Experientia, 23, 699 (1967)SKU(s): CCKS-001A, CCKS-001B, CCKS-001C
Catalog No. 50-210-9107
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.