Learn More
FIREBIRD OPTICS Calcium Fluoride (CaF2) Circular Optical Window- 38x6mm
SDP
Supplier:  FIREBIRD OPTICS CAF238X6
Encompass_Preferred
Calcium Fluoride (CaF2) Circular Optical Window- 38x6mm• Wavelength range: 130nm-9.5µm •Surface quality: 60/40 • Clear aperture: 90% •Diameter tolerance: ±0.1mm •Thickness tolerance: ±0.2mm
Catalog No. NC1953398
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.