Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cafestol from Coffee Beans, MP Biomedicals™

Catalog No. MP219374525 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219374525 25 mg
MP219374550 50 mg
MP219374580 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP219374525 Supplier MP Biomedicals, Inc Supplier No. 0219374525
Add to Cart
Add to Cart

Cafestol from Coffee Beans, White Powder

  • It is a natural extract which induces glutathione S-transferase.
  • It is used to study mechanisms of anti-oxidation related to hydrogen peroxide induced oxidative stress and DNA damage.
  • It is a diterpene molecule present in coffee.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 469-83-0
Molecular Formula C20H28O3
Molecular Weight (g/mol) 316.44
MDL Number MFCD01075769
InChI Key DNJVYWXIDISQRD-HWUKTEKMSA-N
PubChem CID 108052
ChEBI CHEBI:3291
IUPAC Name (1S,4S,12S,13R,16R,17R)-17-(hydroxymethyl)-12-methyl-8-oxapentacyclo[14.2.1.0¹,¹³.0⁴,¹².0⁵,⁹]nonadeca-5(9),6-dien-17-ol
SMILES C[C@@]12CCC3=C(C=CO3)[C@H]1CC[C@@]13C[C@@H](CC[C@@H]21)[C@@](O)(CO)C3

Specifications

Melting Point 156°C to 158°C (literature)
Color White
Density 1.23g/cm3 at 20 (lit.)
Boiling Point 468.59°C (at 760 mmHg (predicted))
Refractive Index 1.60 (Predicted) (lit.)
Quantity 25 mg
Solubility Information Soluble in DMSO (∽5mg/mL), ethanol (∽5mg/mL), DMF (∽12mg/mL), and 1:5 solution of DMF:PBS(pH 7.2) (∽0.02mg/mL). Insoluble in water.
Physical Form Powder
Assay 0.986
RTECSNumber PB9185090
storageNote1 +4°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.