Learn More
Creative Biomart Recombinant Streptococcus pneumoniae type 14 Capsular Polysaccharide Antigen
Supplier:  Creative Biomart CAP244
Encompass_Preferred
Streptococcus pneumoniae type 14 capsular polysaccharide antigen is a vaccine that contains highly purified capsular polysaccharides from the invasive pneumococcal type 14 of Streptococcus pneumoniae. It is an active immunization for intramuscular or subcutaneous injection against pneumococcal disease such as pneumococcal pneumonia and pneumococcal bacteremia.
Catalog No. 50-208-4622
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.