Learn More
Abcam Caged MK 801, NMDA Receptor Blocker, Solid, 10 mg
Description
- Purity: >99%
- Molecular Weight: 460.49 g/mol
- Product Type: Caged, potent, selective, non-competitive, irreversible NMDA receptor open-channel blocker
- Properties: UV-active MK 801 analog; shows anticonvulsant effects in vivo
- Chemical Composition: 100% (4,5-dimethoxy-2-nitrophenyl)methyl (1S,9R)-1-methyl-16-azatetracyclo[7.6.1.02,7.010,15]hexadeca-2,4,6,10,12,14-hexaene-16-carboxylate
- Storage: Store at -20°C
- Shipping: Ambient - Can Ship with Ice
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | NMDA Receptor |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Quantity | 10 mg |
| For Use With (Application) | NMDA receptor blocking |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.