Learn More
Foxx Life Sciences EZFlow Sterile Polyethersulfone (PES) Syringe Filters, 0.22um Sterilizing Membrane, 33mm, 100/PK
Supplier: Foxx Life Sciences 3712415OEM
371-2415-OEM: EZFlow Syringe Filter, 0.22um PES, 33mm, Sterile, 100/pack. EZFlow 13mm, 25mm, and 33mm Syringe Filters are used for prefiltration, clarification, sterilization, or fine particulate removal. These syringe filters are available in a variety of filter membrane materials in both 0.2um and 0.45um pore sizes. The filter membranes are reinforced with polypropylene housing, with a female Luer-lok inlet and a male Luer slip outlet. All materials comply with USP Class VI Standards and are available for both sterile and non-sterile applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.