Learn More
World Precision Instrument Cal-Cell For Endohm 6/12, 12 mm
SDP
Supplier:  World Precision Instrument CALICELL12
Encompass_Preferred
The CaliCell provides a quick and positive way to test STX electrodes, EVOM2s, Endohm, and Ussing chamber. Its membrane makes use of our unique electric current constriction technology to produce resistance readings comparable to those obtained with real cell cultures. The CaliCell does not have to be refrigerated, and can be cleaned and sterilized with alcohol. Readings will not drift over time as long as the unit is kept in good physical condition.
Catalog No. NC2402256
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.