Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKA™ CAL-O-500 Silicon Oil
SDP

Catalog No. 01257900 Shop All IKA Works Products
Change view
Click to view available options
Product Type:
Standard Silicon Oil
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
01-257-900 Standard Silicon Oil
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-257-900 Supplier IKA™ Supplier No. 0025000435
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified standard silicon oil designed to allow you to check the measurement accuracy of your ROTAVISC.

  • Includes certificate
  • For use with a range of ROTAVISC models

Specifications

Product Type Standard Silicon Oil
Viscosity Range 500 cSt Dynamic
Description Period of Validity: 12 months
Temperature Range (Metric) 25°C
Volume (Metric) 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.