Learn More
Diagnostic Biosystems CALBINDIN Calbindin-D
Supplier:  Diagnostic Biosystems MOB55401
Encompass_Preferred
This antibody is specific to a protein of 28 kDa. Calbindin-D-28K is also known as vitamin D-dependent calcium-binding protein or cholecalcin. Calbindin D-28K has a broad tissue distribution; however it exhibits a cell type-specific expression pattern. This antibody reacts specifically with calbindin-D-28K in brain and kidney tissues.
Catalog No. 50-238-5570
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.