Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calbindin D28K Monoclonal Antibody (CB-955)
GREENER_CHOICE

Catalog No. PIMA532993
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
PIMA532993 100 μg
1 options

Catalog No. PIMA532993

Supplier: Invitrogen™ MA532993

Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with 1 mL of PBS buffer to yield a concentration of 100 μg/mL. Positive Control - WB: Rat Brain Tissue, Rat Brain Tissue, Rat Kidney Tissue, Rat Lung Tissue, rat brain tissue, rat kidney tissue, mouse brain tissue. IHC: Mouse Kidney Tissue, Human Intestinal Cancer Tissue, Rat Kidney Tissue, Human Mammary Cancer Tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Calbindin-D-28K (also termed vitamin D-dependent calcium-binding protein, or cholecalcin), is a highly conserved 28kDa calcium binding protein, with broad tissue distribution. It belongs, together with calmodulin, S-100, parvalbumin, troponin C and other proteins, to a family of low molecular weight calcium-binding proteins (CaBPs). These CaBPs have homologous primary structures, which contain conserved polypeptide folds of the EF-hand type for Ca2+ binding. Calbindin-D-28K is found predominantly in subpopulations of central and peripheral nervous system neurons, and in certain epithelial nervous system neurons, and in certain epithelial cells involved in Ca2+ transport such as distal tubular cells and cortical collecting tubules of the kidney, and in enteric neuroendocrine cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calbindin D28K
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CB-955
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation Ascites, 1.2% sodium acetate with 2mg BSA and 0.01mg sodium azide
Gene CALB1
Gene Accession No. P04467, P05937, P07171, P12658
Gene Alias Brain-2; CAB27; CaBP28K; Calb; CALB1; Calb-1; Calbindin; calbindin 1; calbindin 1, (28kD); calbindin 1, 28 kD; calbindin 1, 28kDa; calbindin D28; calbindin-28K; calbindin-D (28k); CalbindinD28K; CB; cerebellar Ca-binding protein spot 35 protein; cerebellar Ca-binding protein, spot 35 protein; D-28K; PCD-29; RTVL-H protein; spot 35 protein; Vitamin D-dependent calcium-binding protein, avian-type
Gene Symbols CALB1
Host Species Mouse
Immunogen Bovine kidney calbindin-D.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12307, 539930, 793, 83839
Target Species Human, Mouse, Rat, Bovine
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.