Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Caspase-9 Inhibitor I

Catalog No. 2187611MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
250 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-876-11MG 1 mg
21-876-1250UG 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-876-11MG Supplier MilliporeSigma™ Supplier No. 2187611MG

The Caspase-9 Inhibitor I controls the biological activity of Caspase-9. This small molecule/inhibitor is primarily used for Cancer applications.

  • Peptide Sequence: Z-Leu-Glu(OMe)-His-Asp(OMe)-CH2F*
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White to Light Brown
Formulation Supplied as a trifluoroacetate salt.
Target Proteins caspase-4, caspase-5, caspase-9
Molecular Formula C32H43FN6O10
Formula Weight 690.7g/mol
Solubility DMSO
Content And Storage Protect from moisture
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.