Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Cdk2 Inhibitor III

Catalog No. 2388031MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-880-31MG 1 mg
23-880-35MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-880-31MG Supplier MilliporeSigma™ Supplier No. 2388031MG
Add to Cart
Add to Cart

The Cdk2 Inhibitor III, also referenced under CAS 199986-75-9, controls the biological activity of Cdk2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins Cdk2/A, Cdk2/E
CAS 199986-75-9
Molecular Formula C20H28N6O3
Formula Weight 400.5g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Purity 98% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.