Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ JNK Inhibitor II

Catalog No. 4201195MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-011-95MG 5 mg
42-011-925MG 25 mg
42-011-950MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 42-011-95MG Supplier MilliporeSigma™ Supplier No. 4201195MG
Add to Cart
Add to Cart

JNK Inhibitor II. SP600125, CAS 129-56-6, is a potent, cell-permeable, selective, and ATP competitive inhibitor of c-Jun N-terminal kinase (JNK; IC50 = 40 nM for JNK-1 & JNK-2 & 90 nM for JNK-3).

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Yellowish Orange
Target Proteins JNK 1, JNK 2, JNK 3
CAS 129-56-6
Molecular Formula C14H8N2O
Formula Weight 220.2g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Purity 98% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.