Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ G-Protein βγ Binding Peptide, mSIRK

Catalog No. 3718181MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-181-81MG 1 mg
37-181-85MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 37-181-81MG Supplier MilliporeSigma™ Supplier No. 3718181MG
Add to Cart
Add to Cart

A cell-permeable, N-myristoylated G-protein βγ selective binding peptide that disrupts the interaction between α and βγ subunits and promotes the α subunit dissociation without stimulating nucleotide exchange.

A cell-permeable, N-myristoylated G-protein βγ selective binding peptide that disrupts the interaction between α and βγ subunits and promotes the α subunit dissociation without stimulating nucleotide exchange. Shown to cause a rapid activation of ERK1/2 (EC50 ∽ 2.5 - 5μM) in rat arterial smooth muscle and Rat2 cells. Also promotes the phosphorylation of JNK and p38 MAP kinase, phospholipase C and causes Ca2+ release from internal stores. Negative control is also available (Cat. No. 371820).
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a trifluoroacetate salt.
Target Proteins N-myristoylated G-protein βγ selective binding
Molecular Formula C93H150N20O25
Formula Weight 1948.3g/mol
Solubility DMSO
Content And Storage Hygroscopic
Form Lyophilized
Quantity 1 mg
Purity 95% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.