Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Chymostatin

Catalog No. 2307905MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-079-05MG 5 mg
23-079-010MG 10 mg
23-079-025MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-079-05MG Supplier MilliporeSigma™ Supplier No. 2307905MG
Add to Cart
Add to Cart

Contains chymostatin A as a major form.

Contains chymostatin A as a major form. Protease inhibitor produced by actinomycetes. Specifically inhibits chymotrypsin (ID50 = 150 ng/ml), papain (ID50 = 7.5μg/ml), and most cysteine proteases.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white with a Pink Cast
Target Proteins chymotrypsin
CAS 9076-44-2
Molecular Formula C25H30N6O5X (X varies from lot to lot)
Solubility DMSO
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.