Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ IL-1R Antagonist

Catalog No. 40761625MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-761-625MG 25 mg
40-761-610MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 40-761-625MG Supplier MilliporeSigma™ Supplier No. 40761625MG
Add to Cart
Add to Cart

The IL-1R Antagonist, also referenced under CAS 566914-00-9, controls the biological activity of IL-1R.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
Target Proteins TIR domain-mediated MyD88/IL1-RI interaction
CAS 566914-00-9
Molecular Formula C18H26N2O2
Formula Weight 302.4g/mol
Solubility DMSO or EtOH
Form Oil
Quantity 25 mg
Purity 95% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.