Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ KT5823

Catalog No. 420321100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-032-1100UG 100 μg
42-032-150UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 42-032-1100UG Supplier MilliporeSigma™ Supplier No. 420321100UG
Add to Cart
Add to Cart

Highly specific, cell-permeable, reversible, and ATP-competitive inhibitor of protein kinase G (Ki = 234 nM).

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins PKG
CAS 126643-37-6
Molecular Formula C29H25N3O5
Formula Weight 495.5g/mol
Solubility DMF or DMSO
Content And Storage Protect from light
Form Solid
Quantity 100 μg
Purity 98% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.