Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Bisindolylmaleimide I

Catalog No. 2032901MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
250 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-329-01MG 1 mg
20-329-0250UG 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 20-329-01MG Supplier MilliporeSigma™ Supplier No. 2032901MG
Add to Cart
Add to Cart

A highly selective, cell-permeable, and reversible protein kinase C (PKC) inhibitor (IC50 = 10 nM) that is structurally similar to staurosporine.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Deep Orange
Target Proteins PKC
CAS 133052-90-1
Molecular Formula C25H24N4O2
Formula Weight 412.5g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Purity 95% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.