Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Lactacystin, Synthetic

Catalog No. 4261001MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
200 μg
400 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-610-01MG 1 mg
42-610-0200UG 200 μg
42-610-0400UG 400 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 42-610-01MG Supplier MilliporeSigma™ Supplier No. 4261001MG
Add to Cart
Add to Cart

Lactacystin, Synthetic, CAS 1258004-00-0, is an irreversible inhibitor of 20S proteasome inhibitor (IC50 = 500 nM). A covalent inhibitor of the chymotrypsin & trypsin-like activities of proteasome.

An irreversible proteasome inhibitor. A Streptomyces metabolite that acts as a highly specific inhibitor of the 20S proteasome (MCP; multicatalytic proteinase complex) (IC50 = 500nM for 20S proteasome). Blocks proteasome activity by targeting the catalytic β-subunit. Acts as a covalent inhibitor of the chymotrypsin- and trypsin-like activities of the proteasome. Induces neurite outgrowth in Neuro 2A mouse neuroblastoma cells and inhibits progression of synchronized Neuro 2A cells and MG-63 human osteosarcoma cells beyond the G1 phase of the cell cycle. Reported to induce apoptosis in human monoblast U937 cells. Also inhibits NF-κB activation (IC50 = 10μM).
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins 20S proteasome (MCP; multicatalytic proteinase complex)
CAS 1258004-00-0
Molecular Formula C15H24N2O7S
Formula Weight 376.4g/mol
Solubility DMSO or H2O
Form Solid
Quantity 1 mg
Purity 99% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.