Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Agar

Catalog No. 121771KG Shop All MilliporeSigma Products
Change view
Click to view available options
Agar; Qty: 1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
12-177-1KG Agar; Qty: 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-177-1KG Supplier MilliporeSigma™ Supplier No. 121771KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suitable for inclusion in culture media.

  • Form: Beige Solid
TRUSTED_SUSTAINABILITY

Specifications

Color Beige
Content And Storage 15°C to 30°C
Form Solid
Product Type Agar
For Use With (Application) Culture media
Packaging Type Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.