Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ LY 294002

Catalog No. 44020225MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-020-225MG 25 mg
44-020-25MG 5 mg
44-020-210MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 44-020-225MG Supplier MilliporeSigma™ Supplier No. 44020225MG
Add to Cart
Add to Cart

LY294002, CAS 154447-36-6, is a cell-permeable, potent, reversible, and specific inhibitor of PI 3-kinase ((IC50 = 1.4 μM). Acts on the ATP-binding site.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Target Proteins Phosphatidylinositol 3-kinase
CAS 154447-36-6
Molecular Formula C19H17NO3
Formula Weight 307.4g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 25 mg
Purity 98% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.