Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ α2-Macroglobulin, Human Plasma

Catalog No. p-7060099 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-125-11MG 1 mg
44-125-110MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 44-125-11MG Supplier MilliporeSigma™ Supplier No. 4412511MG
Add to Cart
Add to Cart

A multifunctional, secreted glycoprotein that acts as a broad range irreversible protease inhibitor that forms a trap around most proteases.

A multifunctional, secreted glycoprotein that acts as a broad range irreversible protease inhibitor that forms a trap around most proteases. α2M is so large a molecule that it tends to remain intravascular. Localized on the surface of peripheral blood lymphocytes. Multifunctional activities include promoting growth of mammalian cells in culture, stimulating regeneration of lymphocytes in irradiated mice, transporting zinc, and acting as proteinase inhibitor that controls the clotting and fibrinolytic system.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 30 mM sodium phosphate buffer, with glycine, pH 7.0.
Target Proteins broad range protease
Formula Weight 725000g/mol
Solubility Aqueous buffers
Form Lyophilized
Quantity 1 mg
Purity 95% by SDS-PAGE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.