Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ MG-132
GSA_VA

Catalog No. 47479010MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-479-010MG 10 mg
47-479-01MG 1 mg
47-479-05MG 5 mg
47-479-020MG 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 47-479-010MG Supplier MilliporeSigma™ Supplier No. 47479010MG
Add to Cart
Add to Cart

A cell-permeable, potent, reversible proteasome inhibitor (Ki = 4 nM).

  • Peptide Sequence: Z-Leu-Leu-Leu-CHO
  • Cell Permeable

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins proteasome
CAS 133407-82-6
Molecular Formula C26H41N3O5
Formula Weight 475.6g/mol
Solubility DMSO or EtOH
Form Solid
Quantity 10 mg
Purity 98% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.