Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Monastrol

Catalog No. 4758791MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-587-91MG 1 mg
47-587-95MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 47-587-91MG Supplier MilliporeSigma™ Supplier No. 4758791MG
Add to Cart
Add to Cart

A potent, cell-permeable, non-tubulin-interacting mitosis inhibitor.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins Mitotic kinesin Eg5
CAS 254753-54-3
Molecular Formula C14H16N2O3S
Formula Weight 292.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Purity 95% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.