Learn More
MilliporeSigma™ Calbiochem™ NG-Monomethyl-L-arginine, Monoacetate Salt
Shop All MilliporeSigma ProductsDescription
- Packaged Under Inert Gas
- Cell Permeable
Specifications
Specifications
| Color | White |
| Target Proteins | eNOS |
| CAS | 53308-83-1 |
| Molecular Formula | C7H16N4O2·CH3CO2H |
| Formula Weight | 248.3g/mol |
| Solubility | MeOH or H2O |
| Content And Storage | Hygroscopic |
| Form | Solid |
| Quantity | 100 mg |
| Purity | 99% by TLC; 99% by HPLC |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.