Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PD 98059

Catalog No. 51300010MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-300-010MG 10 mg
51-300-05MG 5 mg
51-300-025MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 51-300-010MG Supplier MilliporeSigma™ Supplier No. 51300010MG
Add to Cart
Add to Cart

PD 98059, CAS 167869-21-8, is a cell-permeable, selective & reversible inhibitor of MAP Kinase Kinase (MEK). Inhibits MAP Kinase activation and subsequent phosphorylation of MAP Kinase substrates.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Pale Yellow
Target Proteins MAP kinase kinase (MEK)
CAS 167869-21-8
Molecular Formula C16H13NO3
Formula Weight 267.3g/mol
Solubility DMSO or MeOH
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Purity 98% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.