Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Phenylmethylsulfonyl Fluoride

Catalog No. 5233225GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-332-25GM 25 g
52-332-1GM 1 g
52-332-5GM 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 52-332-25GM Supplier MilliporeSigma™ Supplier No. 5233225GM
Add to Cart
Add to Cart

Phenylmethylsulfonyl Fluoride, CAS 329-98-6, is an irreversible inhibitor of serine proteases. It causes sulfonylation of the active-site serine residues.

Irreversible inhibitor of serine proteases. Its mechanism of action is analogous to that of diisopropylfluorophosphate. PMSF causes sulfonylation of the active-site serine residues. Also reported to inhibit internucleosomal DNA fragmentation in immature thymocytes. For a related, more stable inhibitor, see AEBSF (Cat. No. 101500).
TRUSTED_SUSTAINABILITY

Specifications

Color White to Off-white
Target Proteins serine proteases
CAS 329-98-6
Molecular Formula C7H7FO2S
Formula Weight 174.2g/mol
Solubility EtOH, isopropanol, or MeOH
Content And Storage Protect from moisture
Form Crystalline Solid
Quantity 25 g
Purity 99% by GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.