Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Albumin, Human Serum, Fraction V

Catalog No. 12668100GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-668-100GM 100 g
12-668-10GM 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-668-100GM Supplier MilliporeSigma™ Supplier No. 12668100GM
Add to Cart
Add to Cart

Prepared under non-denaturing conditions by modification of Cohn procedure

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 g
Source Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV
Storage Requirements 2° to 8°C
Form Solid
Purity or Quality Grade ≥95% by SDS PAGE

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.