Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Albumin, Human Serum, Non-denatured

Catalog No. 1266541GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-665-41GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-665-41GM Supplier MilliporeSigma™ Supplier No. 1266541GM
Add to Cart
Add to Cart

Suitable for use as Antigen, Culture media for animal cells, Protein standard

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Antigen for Antibody Production, Mammalian Cell Culture, Protein Standard
Quantity 1 g
Source Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.
Storage Requirements 2° to 8°C
Form Lyophilized
Purity or Quality Grade ≥95% by SDS-PAGE

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.